Assay Details
Target Gene Details
Entrez Gene ID: | 5228 |
Gene Name: | placental growth factor |
Gene Aliases: |
D12S1900, PGFL, PIGF, PLGF, PlGF-2, SHGC-10760 |
Location: |
Chr.14:74941830-74955764 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGF | NM_001207012.1 | NP_001193941.1 | ||
| NM_001293643.1 | NP_001280572.1 | |||
| NM_002632.5 | NP_002623.2 | |||
| A18411.1 | CAA01393.1 | |||
| AK309886.1 | ||||
| AK310608.1 | ||||
| AK310886.1 | ||||
| AL543693.3 | ||||
| BC001422.2 | AAH01422.1 | |||
| BC007255.1 | AAH07255.1 | |||
| BC007789.2 | AAH07789.1 | |||
| BT007182.1 | AAP35846.1 | |||
| BX248289.1 | CAD62617.1 | |||
| S72960.1 | AAB30462.2 | |||
| X54936.1 | CAA38698.1 |
More Information
Set Membership: |
|

Genomic Map