Assay Details
Target Gene Details
Entrez Gene ID: | 10243 |
Gene Name: | gephyrin |
Gene Aliases: |
GEPH, GPH, GPHRYN, HKPX1, MOCODC |
Location: |
Chr.14:66507407-67735831 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPHN | XM_017020925.1 | XP_016876414.1 |
Target Gene Details
Entrez Gene ID: | 5283 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class H |
Gene Aliases: |
GPI-H |
Location: |
Chr.14:67589306-67600302 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGH | NM_004569.3 | NP_004560.1 | ||
| XM_011536838.2 | XP_011535140.1 | |||
| XM_017021371.1 | XP_016876860.1 | |||
| XM_017021372.1 | XP_016876861.1 | |||
| AK314108.1 | ||||
| BC004100.2 | AAH04100.1 | |||
| BC071849.1 | AAH71849.1 | |||
| BP244435.1 | ||||
| BT006804.1 | AAP35450.1 | |||
| L19783.1 | AAA03545.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892557 | Chr.14:67469765 - 67613847 on Build GRCh38 | Gain |
|
| nsv564961 | Chr.14:67266144 - 67637063 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs143844247] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map