Assay Details
Target Gene Details
Entrez Gene ID: | 79609 |
Gene Name: | valosin containing protein lysine methyltransferase |
Gene Aliases: |
C14orf138, METTL21D, VCP-KMT |
Location: |
Chr.14:50100456-50118343 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VCPKMT | NM_001040662.1 | NP_001035752.1 | ||
| NM_024558.2 | NP_078834.2 | |||
| NR_049738.1 | ||||
| NR_049739.1 | ||||
| XM_005268060.3 | XP_005268117.1 | |||
| XM_017021640.1 | XP_016877129.1 | |||
| XM_017021641.1 | XP_016877130.1 | |||
| XM_017021642.1 | XP_016877131.1 | |||
| AK023982.1 | BAB14752.1 | |||
| AK129564.1 | BAC85183.1 | |||
| BC143673.1 | ||||
| BC143674.1 | ||||
| BI753530.1 | ||||
| BI767189.1 | ||||
| BX247997.1 | CAD62329.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564835 | Chr.14:50024482 - 50439753 on Build GRCh38 | Gain |
|
| esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map