Assay Details
Target Gene Details
Entrez Gene ID: | 9229 |
Gene Name: | DLG associated protein 1 |
Gene Aliases: |
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1 |
Location: |
Chr.18:3496032-4455307 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Target Gene Details
Entrez Gene ID: | 84777 |
Gene Name: | DLGAP1 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.18:3603737-3610089 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLGAP1-AS2 | NR_119377.1 | |||
| AK091102.1 | 1 | 1184 | ||
| BG830620.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv184n21 | Chr.18:3526777 - 5930980 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map