Assay Details
Target Gene Details
Entrez Gene ID: | 4152 |
Gene Name: | methyl-CpG binding domain protein 1 |
Gene Aliases: |
CXXC3, PCM1, RFT |
Location: |
Chr.18:50266882-50281774 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBD1 | NM_001204139.1 | NP_001191068.1 | ||
| NM_001204141.1 | NP_001191070.1 | |||
| NM_001323951.1 | NP_001310880.1 | |||
| NM_001323953.1 | NP_001310882.1 | |||
| XM_011525998.2 | XP_011524300.1 | |||
| XM_017025754.1 | XP_016881243.1 | |||
| XM_017025755.1 | XP_016881244.1 | |||
| XM_017025761.1 | XP_016881250.1 | |||
| XM_017025768.1 | XP_016881257.1 | |||
| XM_017025772.1 | XP_016881261.1 | |||
| XM_017025775.1 | XP_016881264.1 | |||
| XM_017025777.1 | XP_016881266.1 | |||
| AB209765.1 | BAD93002.1 | |||
| AK291519.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576889 | Chr.18:50227099 - 50287465 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map