Assay Details
Target Gene Details
Entrez Gene ID: | 54814 |
Gene Name: | glutaminyl-peptide cyclotransferase like |
Gene Aliases: |
gQC |
Location: |
Chr.19:45692402-45703990 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QPCTL | NM_001163377.1 | NP_001156849.1 | ||
| NM_017659.3 | NP_060129.2 | |||
| XM_011527048.2 | XP_011525350.1 | |||
| XM_017026900.1 | XP_016882389.1 | |||
| AK000091.1 | BAA90938.1 | |||
| AK172764.1 | BAD18747.1 | |||
| AK222636.1 | BAD96356.1 | |||
| BC011553.1 | AAH11553.1 |
More Information
Set Membership: |
|

Genomic Map