Assay Details
Target Gene Details
Entrez Gene ID: | 148113 |
Gene Name: | cartilage intermediate layer protein 2 |
Gene Aliases: |
CLIP-2 |
Location: |
Chr.19:19538265-19546659 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CILP2 | NM_153221.2 | 1 | 104 | NP_694953.2 |
| AF542080.1 | AAN17826.1 | |||
| AK127735.1 | 1 | 121 | ||
| AK314213.1 | ||||
| DA328640.1 | 1 | 104 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509723 | Chr.19:19517547 - 19595561 on Build GRCh38 | Insertion |
|
| nsv952426 | Chr.19:19525792 - 19601091 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map