Assay Details
Target Gene Details
Entrez Gene ID: | 147923 |
Gene Name: | zinc finger protein 420 |
Gene Aliases: |
APAK |
Location: |
Chr.19:37007815-37130322 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF420 | XM_011526503.2 | XP_011524805.1 | ||
| XM_011526510.2 | XP_011524812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509740 | Chr.19:36970878 - 37041708 on Build GRCh38 | Insertion |
|
| nsv953291 | Chr.19:37007199 - 37016698 on Build GRCh38 | Deletion |
|
| nsv579461 | Chr.19:36834924 - 37256206 on Build GRCh38 | Gain |
|
| esv2762037 | Chr.19:37012997 - 37019882 on Build GRCh38 | Loss |
|
| esv3893201 | Chr.19:36973473 - 37170023 on Build GRCh38 | Gain |
|
| nsv1064851 | Chr.19:36419357 - 37198819 on Build GRCh38 | Loss |
|
| nsv833818 | Chr.19:36936906 - 37111898 on Build GRCh38 | Gain |
|
| nsv961214 | Chr.19:37003797 - 37015325 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs192230317] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map