Assay Details
Target Gene Details
Entrez Gene ID: | 388512 |
Gene Name: | C-type lectin domain family 17 member A |
Gene Aliases: |
- |
Location: |
Chr.19:14580777-14612035 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC17A | XM_017026788.1 | 1 | 136 | XP_016882277.1 |
More Information
Set Membership: |
|

Genomic Map