Assay Details
Target Gene Details
Entrez Gene ID: | 6449 |
Gene Name: | small glutamine rich tetratricopeptide repeat containing alpha |
Gene Aliases: |
SGT, alphaSGT, hSGT |
Location: |
Chr.19:2754714-2783356 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGTA | NM_003021.3 | NP_003012.1 | ||
| XM_011528178.2 | XP_011526480.1 | |||
| AF368279.1 | AAP29457.1 | |||
| AF408399.1 | AAL01051.1 | |||
| AJ133129.1 | CAB39725.1 | |||
| AJ223828.1 | CAA11565.1 | |||
| AK293535.1 | ||||
| AL050156.1 | CAB43297.2 | |||
| BC000390.2 | AAH00390.1 | |||
| BC002989.1 | AAH02989.2 | |||
| BC005165.2 | AAH05165.1 | |||
| BC008885.2 | AAH08885.1 | |||
| CR533517.1 | CAG38548.1 | |||
| CR542282.1 | CAG47077.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509714 | Chr.19:2519629 - 2794403 on Build GRCh38 | Insertion |
|
| nsv828397 | Chr.19:2688987 - 2800506 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map