Assay Details
Target Gene Details
Entrez Gene ID: | 100506068 |
Gene Name: | DACT3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:46660364-46677456 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DACT3-AS1 | NR_040041.1 | 1 | 680 | |
| NR_040042.1 | ||||
| AW269981.1 | ||||
| BX116982.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833849 | Chr.19:46558831 - 46703060 on Build GRCh38 | Loss |
|

Genomic Map