Assay Details
Target Gene Details
Entrez Gene ID: | 105372412 |
Gene Name: | phospholipase A2 inhibitor and Ly6/PLAUR domain-containing protein-like |
Gene Aliases: |
- |
Location: |
Chr.19:43687557-43700540 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105372412 | XM_017027562.1 | XP_016883051.1 | ||
| XM_017027563.1 | XP_016883052.1 | |||
| XM_017027564.1 | XP_016883053.1 | |||
| XM_017027565.1 | XP_016883054.1 | |||
| XM_017027566.1 | XP_016883055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2503 | Chr.19:43690920 - 43723211 on Build GRCh38 | Insertion |
|
| esv3644474 | Chr.19:43687730 - 43702516 on Build GRCh38 | Gain |
|
| nsv833839 | Chr.19:43595661 - 43768147 on Build GRCh38 | Loss |
|
| esv3893220 | Chr.19:43658802 - 43706225 on Build GRCh38 | Gain |
|
| nsv819607 | Chr.19:43692441 - 43697048 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map