Assay Details
Target Gene Details
Entrez Gene ID: | 4782 |
Gene Name: | nuclear factor I C |
Gene Aliases: |
CTF, CTF5, NF-I, NFI |
Location: |
Chr.19:3359563-3469217 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFIC | NM_001245002.1 | NP_001231931.1 | ||
| NM_001245004.1 | NP_001231933.1 | |||
| NM_001245005.1 | NP_001231934.1 | |||
| NM_005597.3 | NP_005588.2 | |||
| NM_205843.2 | NP_995315.1 | |||
| XM_005259564.3 | XP_005259621.1 | |||
| XM_006722759.2 | XP_006722822.2 | |||
| XM_017026834.1 | XP_016882323.1 | |||
| XM_017026835.1 | XP_016882324.1 | |||
| XM_017026836.1 | XP_016882325.1 | |||
| AK289885.1 | ||||
| AK297825.1 | ||||
| AK297867.1 | ||||
| AK304816.1 | ||||
| BC012120.1 | AAH12120.1 | |||
| BF763988.1 | ||||
| BP279194.1 | ||||
| BQ894051.1 | ||||
| X12492.1 | CAA31012.1 | |||
| X92857.1 | CAA63440.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509717 | Chr.19:3271020 - 3392050 on Build GRCh38 | Insertion |
|
| nsv1058546 | Chr.19:3321667 - 3369170 on Build GRCh38 | Gain |
|
| nsv953948 | Chr.19:3171703 - 3700002 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map