Assay Details
Target Gene Details
Entrez Gene ID: | 10856 |
Gene Name: | RuvB like AAA ATPase 2 |
Gene Aliases: |
CGI-46, ECP-51, ECP51, INO80J, REPTIN, RVB2, TAP54-beta, TIH2, TIP48, TIP49B |
Location: |
Chr.19:48993448-49015995 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RUVBL2 | NM_001321190.1 | NP_001308119.1 | ||
| NM_001321191.1 | NP_001308120.1 | |||
| NM_006666.2 | NP_006657.1 | |||
| NR_135578.1 | ||||
| XM_011526330.1 | XP_011524632.1 | |||
| AB024301.1 | BAA76708.1 | |||
| AF124607.1 | AAF87087.1 | |||
| AF151804.1 | AAD34041.1 | |||
| AF155138.1 | AAD38073.1 | |||
| AK027762.1 | ||||
| AK057498.1 | ||||
| AK074542.1 | BAC11048.1 | |||
| AK301344.1 | ||||
| BC000428.2 | AAH00428.1 | |||
| BC004531.2 | AAH04531.1 | |||
| BC008355.1 | AAH08355.1 | |||
| Y18417.1 | CAB46270.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| dgv3618n100 | Chr.19:48948950 - 49021211 on Build GRCh38 | Gain |
|
| nsv833857 | Chr.19:48914476 - 49146320 on Build GRCh38 | Loss |
|
| nsv953598 | Chr.19:48957644 - 49085143 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map