Assay Details
Target Gene Details
Entrez Gene ID: | 3783 |
Gene Name: | potassium calcium-activated channel subfamily N member 4 |
Gene Aliases: |
DHS2, IK, IK1, IKCA1, KCA4, KCa3.1, SK4, hIKCa1, hKCa4, hSK4 |
Location: |
Chr.19:43766533-43782139 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNN4 | NM_002250.2 | NP_002241.1 | ||
| XM_005258882.2 | XP_005258939.1 | |||
| XM_005258883.2 | XP_005258940.1 | |||
| AB128983.1 | ||||
| AB128984.1 | ||||
| AB128985.1 | ||||
| AF000972.1 | AAB82739.1 | |||
| AF022150.1 | AAC23541.1 | |||
| AF022797.1 | AAC51913.1 | |||
| AF033021.1 | AAC36804.1 | |||
| AF053403.1 | AAC35281.1 | |||
| AF395661.1 | AAK81862.1 | |||
| BC015337.1 | AAH15337.1 | |||
| BT007426.1 | AAP36094.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509745 | Chr.19:43746288 - 43808495 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map