Assay Details
Target Gene Details
Entrez Gene ID: | 4858 |
Gene Name: | NOVA alternative splicing regulator 2 |
Gene Aliases: |
ANOVA, NOVA3 |
Location: |
Chr.19:45933734-45973399 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOVA2 | NM_002516.3 | NP_002507.1 | ||
| XM_006723230.3 | XP_006723293.1 | |||
| XM_017026839.1 | XP_016882328.1 | |||
| AF083898.1 | AAC72355.1 | |||
| U70477.1 | AAB88661.1 |
More Information
Set Membership: |
|

Genomic Map