Assay Details
Target Gene Details
Entrez Gene ID: | 3337 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B1 |
Gene Aliases: |
HSPF1, Hdj1, Hsp40, RSPH16B, Sis1 |
Location: |
Chr.19:14514764-14529821 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB1 | NM_001300914.1 | NP_001287843.1 | ||
| NM_001313964.1 | NP_001300893.1 | |||
| NM_006145.2 | NP_006136.1 | |||
| XM_006722733.1 | XP_006722796.1 | |||
| XM_006722734.3 | XP_006722797.1 | |||
| XM_011527956.2 | XP_011526258.1 | |||
| AK296609.1 | ||||
| AK301817.1 | ||||
| AK312624.1 | ||||
| BC002352.2 | AAH02352.1 | |||
| BC019827.1 | AAH19827.1 | |||
| CR536485.1 | CAG38724.1 | |||
| CR541677.1 | CAG46478.1 | |||
| D49547.1 | BAA08495.1 | |||
| DA377896.1 | ||||
| DB454482.1 | ||||
| HY032238.1 | ||||
| HY042250.1 | ||||
| X62421.1 | CAA44287.1 |
More Information
Set Membership: |
|

Genomic Map