Assay Details
Target Gene Details
Entrez Gene ID: | 3949 |
Gene Name: | low density lipoprotein receptor |
Gene Aliases: |
FH, FHC, LDLCQ2 |
Location: |
Chr.19:11089362-11133830 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LDLR | NM_000527.4 | NP_000518.1 | ||
| NM_001195798.1 | NP_001182727.1 | |||
| NM_001195799.1 | NP_001182728.1 | |||
| NM_001195800.1 | NP_001182729.1 | |||
| NM_001195803.1 | NP_001182732.1 | |||
| XM_011528010.1 | XP_011526312.1 | |||
| AB209409.1 | BAD92646.1 | |||
| AK295612.1 | ||||
| AK296312.1 | ||||
| AK299038.1 | ||||
| AK300313.1 | ||||
| AY114155.1 | AAM56036.1 | |||
| BC014514.1 | AAH14514.1 | |||
| BT007361.1 | AAP36025.1 | |||
| BX648281.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833751 | Chr.19:11006139 - 11164225 on Build GRCh38 | Loss |
|
| nsv470126 | Chr.19:10994932 - 11116926 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map