Assay Details
Target Gene Details
Entrez Gene ID: | 3337 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B1 |
Gene Aliases: |
HSPF1, Hdj1, Hsp40, RSPH16B, Sis1 |
Location: |
Chr.19:14514764-14529821 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB1 | NM_001313964.1 | NP_001300893.1 | ||
| XM_006722733.1 | XP_006722796.1 | |||
| XM_006722734.3 | XP_006722797.1 | |||
| XM_011527956.2 | XP_011526258.1 | |||
| AK301817.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115330843] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map