Assay Details
Target Gene Details
Entrez Gene ID: | 5141 |
Gene Name: | phosphodiesterase 4A |
Gene Aliases: |
DPDE2, PDE4, PDE46 |
Location: |
Chr.19:10416773-10469631 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE4A | NM_001111307.1 | NP_001104777.1 | ||
| NM_001243121.1 | NP_001230050.1 | |||
| XM_011528054.1 | XP_011526356.1 | |||
| XM_011528055.2 | XP_011526357.1 | |||
| XM_017026865.1 | XP_016882354.1 | |||
| AY593872.1 | AAT00628.1 | |||
| L20965.1 | AAA03588.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521918 | Chr.19:10426592 - 10431934 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map