Assay Details
Target Gene Details
Entrez Gene ID: | 10298 |
Gene Name: | p21 (RAC1) activated kinase 4 |
Gene Aliases: |
- |
Location: |
Chr.19:39125780-39179406 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAK4 | NM_001014831.2 | NP_001014831.1 | ||
| NM_001014832.1 | NP_001014832.1 | |||
| NM_001014834.2 | NP_001014834.1 | |||
| NM_001014835.1 | NP_001014835.1 | |||
| NM_005884.3 | NP_005875.1 | |||
| XM_011526316.1 | XP_011524618.1 | |||
| XM_011526320.1 | XP_011524622.1 | |||
| AB032968.1 | ||||
| AF005046.1 | AAD01210.1 | |||
| AK291839.1 | ||||
| AK293517.1 | ||||
| AK294586.1 | ||||
| AK300632.1 | ||||
| AK304651.1 | ||||
| AL834236.1 | CAD38914.2 | |||
| AX775863.1 | CAE11631.1 | |||
| AX775865.1 | CAE11632.1 | |||
| AX775867.1 | CAE11633.1 | |||
| AX775869.1 | CAE11634.1 | |||
| AX775871.1 | CAE11635.1 | |||
| BC002921.2 | AAH02921.1 | |||
| BC011368.1 | AAH11368.1 | |||
| BC025282.1 | AAH25282.1 | |||
| BC034511.1 | AAH34511.1 | |||
| BG773480.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833829 | Chr.19:39079041 - 39240137 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115728047] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map