Assay Details
Target Gene Details
Entrez Gene ID: | 5518 |
Gene Name: | protein phosphatase 2 scaffold subunit Aalpha |
Gene Aliases: |
MRD36, PP2A-Aalpha, PP2AAALPHA, PR65A |
Location: |
Chr.19:52189802-52226425 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R1A | NM_014225.5 | NP_055040.2 | ||
| NR_033500.1 | ||||
| AK091387.1 | BAC03652.1 | |||
| AK130000.1 | ||||
| AK290593.1 | ||||
| AK291932.1 | ||||
| AK293174.1 | ||||
| AK293493.1 | ||||
| AK299012.1 | ||||
| AK307743.1 | ||||
| AK315700.1 | ||||
| AU128747.1 | ||||
| BC001537.2 | AAH01537.1 | |||
| CR450340.1 | CAG29336.1 | |||
| DB246691.1 | ||||
| DC398468.1 | ||||
| J02902.1 | AAA36399.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059417 | Chr.19:52159127 - 52200548 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map