Assay Details
Target Gene Details
Entrez Gene ID: | 90273 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 21 |
Gene Aliases: |
CEACAM3, R29124_1 |
Location: |
Chr.19:41549518-41586844 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEACAM21 | NM_001288773.2 | NP_001275702.2 | ||
| XM_011527450.1 | XP_011525752.1 | |||
| XM_011527452.1 | XP_011525754.1 | |||
| XM_017027433.1 | XP_016882922.1 | |||
| XM_017027436.1 | XP_016882925.1 | |||
| AK023602.1 | ||||
| BI913326.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6455n54 | Chr.19:41553480 - 41555814 on Build GRCh38 | Loss |
|
| nsv579603 | Chr.19:41551655 - 41555761 on Build GRCh38 | Loss |
|
| esv2668558 | Chr.19:41552944 - 41555769 on Build GRCh38 | Deletion |
|
| esv3556412 | Chr.19:41552653 - 41555772 on Build GRCh38 | Deletion |
|
| esv3583419 | Chr.19:41554347 - 41557001 on Build GRCh38 | Loss |
|
| esv3644387 | Chr.19:41552950 - 41555787 on Build GRCh38 | Loss |
|
| dgv6456n54 | Chr.19:41554997 - 41555814 on Build GRCh38 | Loss |
|
| dgv3549n100 | Chr.19:41550966 - 41671232 on Build GRCh38 | Gain |
|
| nsv442463 | Chr.19:41553476 - 41555933 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map