Assay Details
Target Gene Details
Entrez Gene ID: | 57794 |
Gene Name: | SURP and G-patch domain containing 1 |
Gene Aliases: |
F23858, RBP, SF4 |
Location: |
Chr.19:19276513-19320512 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUGP1 | NM_172231.3 | NP_757386.2 | ||
| XM_005260002.2 | XP_005260059.1 | |||
| XM_011528155.2 | XP_011526457.1 | |||
| AF521128.1 | AAN77123.1 | |||
| AK096152.1 | ||||
| AK292085.1 | ||||
| AK294618.1 | ||||
| AK301119.1 | ||||
| AY072916.1 | AAL68960.1 | |||
| AY072917.1 | AAL68961.1 | |||
| BC063784.1 | AAH63784.1 | |||
| BC119728.1 | ||||
| BC119729.1 | ||||
| BC142988.1 | ||||
| CB960848.1 | ||||
| DC363162.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76944848] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map