Assay Details
Target Gene Details
Entrez Gene ID: | 100506012 |
Gene Name: | PPP5 tetratricopeptide repeat domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.19:46480788-46601200 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP5D1 | NM_001205281.1 | NP_001192210.1 | ||
| BM718195.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583443 | Chr.19:46511536 - 46520153 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map