Assay Details
Target Gene Details
Entrez Gene ID: | 344 |
Gene Name: | apolipoprotein C2 |
Gene Aliases: |
APO-CII, APOC-II |
Location: |
Chr.19:44945982-44949565 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOC2 | NM_000483.4 | NP_000474.2 | ||
| AA883870.1 | ||||
| AV694494.1 | ||||
| BC005348.2 | AAH05348.3 | |||
| BC065270.1 | AAH65270.1 | |||
| BT006708.1 | AAP35354.1 | |||
| BU596546.1 | ||||
| KU177908.1 | ||||
| KU177909.1 | ||||
| M29844.1 | AAA51743.1 | |||
| X00568.1 | CAA25234.1 |
Target Gene Details
Entrez Gene ID: | 100533990 |
Gene Name: | APOC4-APOC2 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.19:44942238-44949565 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOC4-APOC2 | NR_037932.1 | |||
| AK126257.1 |
More Information
Set Membership: |
|

Genomic Map