Assay Details
Target Gene Details
Entrez Gene ID: | 91662 |
Gene Name: | NLR family pyrin domain containing 12 |
Gene Aliases: |
CLR19.3, FCAS2, NALP12, PAN6, PYPAF7, RNO, RNO2 |
Location: |
Chr.19:53793584-53824403 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064558 | Chr.19:53773468 - 53957491 on Build GRCh38 | Gain |
|
| esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map