Assay Details
Target Gene Details
Entrez Gene ID: | 105372441 |
Gene Name: | uncharacterized LOC105372441 |
Gene Aliases: |
KLK3e |
Location: |
Chr.19:50830530-50851143 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105372441 | NR_131203.1 | |||
| NR_131205.1 | ||||
| AA565890.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3583464 | Chr.19:50818135 - 50842312 on Build GRCh38 | Loss |
|
| nsv2531 | Chr.19:50830372 - 50875341 on Build GRCh38 | Deletion |
|
| dgv1058e212 | Chr.19:50828652 - 50842312 on Build GRCh38 | Loss |
|

Genomic Map