Assay Details
Target Gene Details
Entrez Gene ID: | 9524 |
Gene Name: | trans-2,3-enoyl-CoA reductase |
Gene Aliases: |
GPSN2, MRT14, SC2, TER |
Location: |
Chr.19:14527760-14565980 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TECR | NM_001321170.1 | NP_001308099.1 | ||
| NM_138501.5 | NP_612510.1 | |||
| NR_038103.1 | ||||
| NR_038104.1 | ||||
| XM_006722945.1 | XP_006723008.1 | |||
| XM_011528442.2 | XP_011526744.1 | |||
| AF038958.1 | AAC39872.1 | |||
| AF038959.1 | AAC39873.1 | |||
| AF222742.1 | AAF32373.1 | |||
| AK001416.1 | ||||
| AK094087.1 | ||||
| AK299134.1 | ||||
| AK315412.1 | ||||
| BC002425.1 | AAH02425.1 | |||
| BC005952.1 | AAH05952.1 | |||
| BC007801.2 | AAH07801.1 | |||
| BC013881.2 | AAH13881.1 | |||
| BG705196.1 | ||||
| DB491783.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3643744 | Chr.19:14548018 - 14555305 on Build GRCh38 | Loss |
|
| esv3643746 | Chr.19:14550217 - 14554086 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map