Assay Details
Target Gene Details
Entrez Gene ID: | 147923 |
Gene Name: | zinc finger protein 420 |
Gene Aliases: |
APAK |
Location: |
Chr.19:37007815-37130322 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF420 | XM_006723038.3 | XP_006723101.1 | ||
| XM_006723039.3 | XP_006723102.1 | |||
| XM_011526503.2 | XP_011524805.1 | |||
| XM_011526504.2 | XP_011524806.1 | |||
| XM_011526505.2 | XP_011524807.1 | |||
| XM_011526507.2 | XP_011524809.1 | |||
| XM_011526508.2 | XP_011524810.1 | |||
| XM_011526510.2 | XP_011524812.1 | |||
| XM_011526511.2 | XP_011524813.1 | |||
| XM_017026336.1 | XP_016881825.1 | |||
| XM_017026337.1 | XP_016881826.1 | |||
| XM_017026339.1 | XP_016881828.1 | |||
| XM_017026340.1 | XP_016881829.1 | |||
| AK056753.1 | BAB71272.1 | |||
| AK315728.1 | ||||
| BC026210.2 | AAH26210.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv579461 | Chr.19:36834924 - 37256206 on Build GRCh38 | Gain |
|
| esv3893201 | Chr.19:36973473 - 37170023 on Build GRCh38 | Gain |
|
| nsv1064851 | Chr.19:36419357 - 37198819 on Build GRCh38 | Loss |
|
| nsv833818 | Chr.19:36936906 - 37111898 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map