Assay Details
Target Gene Details
Entrez Gene ID: | 55101 |
Gene Name: | ATP5S like |
Gene Aliases: |
- |
Location: |
Chr.19:41431318-41447822 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5SL | NM_001167867.1 | NP_001161339.1 | ||
| NM_001167868.1 | NP_001161340.1 | |||
| NM_001167869.1 | NP_001161341.1 | |||
| NM_001167870.1 | NP_001161342.1 | |||
| NM_001167871.1 | NP_001161343.1 | |||
| NM_001320838.1 | NP_001307767.1 | |||
| NM_001320839.1 | NP_001307768.1 | |||
| NM_001320840.1 | NP_001307769.1 | |||
| NM_001320841.1 | NP_001307770.1 | |||
| NM_001320844.1 | NP_001307773.1 | |||
| NM_018035.2 | NP_060505.2 | |||
| NR_030765.1 | ||||
| NR_135476.1 | ||||
| NR_135477.1 | ||||
| NR_135478.1 | ||||
| XM_011527065.2 | XP_011525367.1 | |||
| AK001103.1 | BAA91503.1 | |||
| AK055565.1 | 1 | 1183 | ||
| AK123631.1 | ||||
| AK293132.1 | ||||
| AK294249.1 | ||||
| AK297698.1 | ||||
| AK298197.1 | ||||
| AK301940.1 | ||||
| AK316313.1 | ||||
| BC013323.1 | AAH13323.1 | |||
| BQ072604.1 | ||||
| DA057313.1 | ||||
| DA075924.1 | ||||
| DA540661.1 | ||||
| DA631852.1 | ||||
| DA703197.1 |

Genomic Map