Assay Details
Target Gene Details
Entrez Gene ID: | 653677 |
Gene Name: | secretory blood group 1, pseudogene |
Gene Aliases: |
SEC1 |
Location: |
Chr.19:48638039-48682245 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC1P | NR_004401.2 | 5 | 1750 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| esv3644596 | Chr.19:48680288 - 48703254 on Build GRCh38 | Loss |
|
| esv2664951 | Chr.19:48680400 - 48703346 on Build GRCh38 | Deletion |
|
| nsv579904 | Chr.19:48680448 - 48699602 on Build GRCh38 | Loss |
|
| nsv521477 | Chr.19:48673773 - 48693829 on Build GRCh38 | Loss |
|

Genomic Map