Assay Details
Target Gene Details
Entrez Gene ID: | 51073 |
Gene Name: | mitochondrial ribosomal protein L4 |
Gene Aliases: |
CGI-28, L4mt |
Location: |
Chr.19:10251964-10260060 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL4 | NM_015956.2 | NP_057040.2 | ||
| NM_146387.1 | NP_666499.1 | |||
| NM_146388.1 | NP_666500.1 | |||
| XM_011528045.2 | XP_011526347.1 | |||
| AB049635.1 | BAB40840.1 | |||
| AF132962.1 | AAD27737.1 | |||
| AY007152.1 | AAG02005.1 | |||
| BC000756.2 | AAH00756.1 | |||
| BC009858.2 | AAH09858.1 | |||
| BG325905.1 | ||||
| BG422689.1 | ||||
| BG758801.1 | ||||
| BM550378.1 |
More Information
Set Membership: |
|

Genomic Map