Assay Details
Target Gene Details
Entrez Gene ID: | 5518 |
Gene Name: | protein phosphatase 2 scaffold subunit Aalpha |
Gene Aliases: |
MRD36, PP2A-Aalpha, PP2AAALPHA, PR65A |
Location: |
Chr.19:52189802-52226425 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R1A | NM_014225.5 | NP_055040.2 | ||
| NR_033500.1 | ||||
| DC398468.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059417 | Chr.19:52159127 - 52200548 on Build GRCh38 | Gain |
|
| nsv580007 | Chr.19:52157026 - 52190024 on Build GRCh38 | Gain |
|
| esv22131 | Chr.19:52189302 - 52192962 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map