Assay Details
Target Gene Details
Entrez Gene ID: | 388512 |
Gene Name: | C-type lectin domain family 17 member A |
Gene Aliases: |
- |
Location: |
Chr.19:14580777-14612035 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC17A | NM_001204118.1 | NP_001191047.1 | ||
| NM_207390.3 | NP_997273.3 | |||
| NR_109784.1 | ||||
| NR_109785.1 | ||||
| XM_017026785.1 | XP_016882274.1 | |||
| XM_017026786.1 | XP_016882275.1 | |||
| XM_017026787.1 | XP_016882276.1 | |||
| XM_017026788.1 | XP_016882277.1 | |||
| XM_017026789.1 | XP_016882278.1 | |||
| XM_017026790.1 | XP_016882279.1 | |||
| XM_017026791.1 | XP_016882280.1 | |||
| XM_017026792.1 | XP_016882281.1 | |||
| XM_017026793.1 | XP_016882282.1 | |||
| XM_017026794.1 | XP_016882283.1 | |||
| BC140848.1 | ||||
| BC144665.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs117570752] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map