Assay Details
Target Gene Details
Entrez Gene ID: | 4605 |
Gene Name: | MYB proto-oncogene like 2 |
Gene Aliases: |
B-MYB, BMYB |
Location: |
Chr.20:43667019-43716496 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYBL2 | NM_001278610.1 | NP_001265539.1 | ||
| NM_002466.3 | NP_002457.1 | |||
| AK223482.1 | BAD97202.1 | |||
| AK303249.1 | ||||
| AK314791.1 | ||||
| BC007585.1 | AAH07585.1 | |||
| BC053555.1 | AAH53555.1 | |||
| DC395847.1 | ||||
| X13293.1 | CAA31655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss |
|
| esv3645886 | Chr.20:43653014 - 43756886 on Build GRCh38 | Gain |
|
| nsv3385 | Chr.20:43641073 - 43676907 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140943696] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map