Assay Details
Target Gene Details
Entrez Gene ID: | 100505826 |
Gene Name: | STK4 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.20:44963794-44966458 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK4-AS1 | NR_038341.1 | |||
| AK057198.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|

Genomic Map