Assay Details
Target Gene Details
Entrez Gene ID: | 128869 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class U |
Gene Aliases: |
CDC91L1, GAB1 |
Location: |
Chr.20:34560542-34677285 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGU | NM_080476.4 | NP_536724.1 | ||
| XM_011528542.2 | XP_011526844.1 | |||
| XM_017027664.1 | XP_016883153.1 | |||
| AB086842.1 | BAC53626.1 | |||
| AK075507.1 | BAC11660.1 | |||
| AK301900.1 | ||||
| AK308627.1 | ||||
| AY339061.1 | AAQ18022.1 | |||
| AY358816.1 | AAQ89175.1 | |||
| AY422169.1 | AAR23798.1 | |||
| BC030512.1 | AAH30512.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75345986] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map