Assay Details
Target Gene Details
Entrez Gene ID: | 55816 |
Gene Name: | docking protein 5 |
Gene Aliases: |
C20orf180, IRS-6, IRS6 |
Location: |
Chr.20:54475472-54651171 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOK5 | NM_001294161.1 | NP_001281090.1 | ||
| NM_018431.4 | NP_060901.2 | |||
| NM_177959.2 | NP_808874.1 | |||
| XM_011528904.1 | XP_011527206.1 | |||
| AF132732.1 | AAF66443.1 | |||
| AF466368.1 | AAL74194.1 | |||
| AI822030.1 | ||||
| AK313222.1 | ||||
| BC008992.1 | AAH08992.1 | |||
| BI758267.1 | ||||
| DA161365.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv586224 | Chr.20:53858311 - 54662951 on Build GRCh38 | Gain |
|
| esv3646138 | Chr.20:54489843 - 54491257 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map