Assay Details
Target Gene Details
Entrez Gene ID: | 2280 |
Gene Name: | FK506 binding protein 1A |
Gene Aliases: |
FKBP-12, FKBP-1A, FKBP1, FKBP12, PKC12, PKCI2, PPIASE |
Location: |
Chr.20:1368977-1393172 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP1A | NM_000801.4 | NP_000792.1 | ||
| NM_001199786.1 | NP_001186715.1 | |||
| NM_054014.3 | NP_463460.1 | |||
| AB241123.1 | ||||
| AB241124.1 | ||||
| AB241125.1 | ||||
| BC001925.1 | ||||
| BC005147.2 | AAH05147.1 | |||
| BC119732.1 | ||||
| BC119733.1 | ||||
| BT007066.1 | AAP35729.1 | |||
| CN426120.1 | ||||
| CR407613.1 | CAG28541.1 | |||
| CR542168.1 | CAG46965.1 | |||
| CX870892.1 | ||||
| M34539.1 | AAA35844.1 | |||
| X52220.1 | CAA36462.1 |
Target Gene Details
Entrez Gene ID: | 100528031 |
Gene Name: | FKBP1A-SDCBP2 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.20:1309909-1393172 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP1A-SDCBP2 | NR_037661.1 | |||
| AK309026.1 | ||||
| BM925547.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522008 | Chr.20:1359746 - 1657932 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map