Assay Details
Target Gene Details
| Entrez Gene ID: | 9139 | 
| Gene Name: | CBFA2/RUNX1 translocation partner 2 | 
| Gene Aliases: | EHT, MTGR1, ZMYND3, p85 | 
| Location: | Chr.20:33490068-33650031 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| CBFA2T2 | NM_001032999.2 | NP_001028171.1 | ||
| XM_017028122.1 | XP_016883611.1 | |||
| AK294168.1 | ||||
| AK307887.1 | ||||
| CB998035.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv1064413 | Chr.20:33348381 - 33599978 on Build GRCh38 | Gain |  CDK5RAP1  SNTA1  CBFA2T2 | 
| dgv4302n100 | Chr.20:33413503 - 33558912 on Build GRCh38 | Loss |  SNTA1  CBFA2T2 | 
| nsv522690 | Chr.20:33427230 - 33552481 on Build GRCh38 | Loss |  SNTA1  CBFA2T2 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top