Assay Details
Target Gene Details
Entrez Gene ID: | 100 |
Gene Name: | adenosine deaminase |
Gene Aliases: |
- |
Location: |
Chr.20:44619519-44651758 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADA | NM_000022.3 | NP_000013.2 | ||
| NM_001322050.1 | NP_001308979.1 | |||
| NM_001322051.1 | NP_001308980.1 | |||
| NR_136160.1 | ||||
| AK223397.1 | BAD97117.1 | |||
| BC007678.2 | AAH07678.1 | |||
| BC040226.1 | AAH40226.1 | |||
| BI601734.1 | ||||
| CD014053.1 | ||||
| CD014056.1 | ||||
| HY102642.1 | ||||
| KJ534760.1 | ||||
| KU177892.1 | ||||
| KU177893.1 | ||||
| KU177894.1 | ||||
| KU177895.1 | ||||
| X02994.1 | CAA26734.1 |
More Information
Set Membership: |
|

Genomic Map