Assay Details
Target Gene Details
Entrez Gene ID: | 26039 |
Gene Name: | SS18L1, nBAF chromatin remodeling complex subunit |
Gene Aliases: |
CREST, LP2261 |
Location: |
Chr.20:62143720-62182510 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SS18L1 | NM_001301778.1 | NP_001288707.1 | ||
| NM_198935.2 | NP_945173.1 | |||
| NR_125980.1 | ||||
| NR_125981.1 | ||||
| XM_005260389.3 | XP_005260446.1 | |||
| XM_005260391.1 | XP_005260448.1 | |||
| XM_017027784.1 | XP_016883273.1 | |||
| AB014593.1 | ||||
| AK125656.1 | ||||
| AK291485.1 | ||||
| AK299881.1 | ||||
| AK308765.1 | ||||
| BX640848.1 | CAE45918.1 | |||
| DC375156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509782 | Chr.20:62125636 - 62180328 on Build GRCh38 | Insertion |
|
| nsv586481 | Chr.20:62143364 - 62158583 on Build GRCh38 | Gain |
|
| nsv3440 | Chr.20:62137705 - 62169472 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78015034] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map