Assay Details
Target Gene Details
Entrez Gene ID: | 10616 |
Gene Name: | RANBP2-type and C3HC4-type zinc finger containing 1 |
Gene Aliases: |
C20orf18, HOIL-1, HOIL1, PBMEI, PGBM1, RBCK2, RNF54, UBCE7IP3, XAP3, XAP4, ZRANB4 |
Location: |
Chr.20:408050-432139 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBCK1 | NM_001323956.1 | NP_001310885.1 | ||
| NM_001323958.1 | NP_001310887.1 | |||
| NM_001323960.1 | NP_001310889.1 | |||
| NM_006462.5 | NP_006453.1 | |||
| NM_031229.3 | NP_112506.2 | |||
| NR_136659.1 | ||||
| XM_005260645.1 | XP_005260702.1 | |||
| XM_011529137.1 | XP_011527439.1 | |||
| XM_011529138.1 | XP_011527440.1 | |||
| XM_011529139.2 | XP_011527441.1 | |||
| XM_011529140.1 | XP_011527442.1 | |||
| XM_017027594.1 | XP_016883083.1 | |||
| AB107766.1 | BAC75409.1 | |||
| AK297524.1 | ||||
| AK303357.1 | ||||
| AK309414.1 | ||||
| BC015219.2 | AAH15219.2 | |||
| U67322.1 | AAD00162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644953 | Chr.20:397290 - 502310 on Build GRCh38 | Gain |
|
| esv3644952 | Chr.20:386013 - 1119183 on Build GRCh38 | Gain |
|
| nsv1056266 | Chr.20:401152 - 522755 on Build GRCh38 | Gain |
|
| esv3644956 | Chr.20:405365 - 446917 on Build GRCh38 | Gain |
|
| nsv510777 | Chr.20:366849 - 416704 on Build GRCh38 | Deletion |
|
| nsv458841 | Chr.20:380145 - 422066 on Build GRCh38 | Loss |
|
| dgv4219n100 | Chr.20:80652 - 461103 on Build GRCh38 | Gain |
|
| esv3893263 | Chr.20:216521 - 434561 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map