Assay Details
Target Gene Details
Entrez Gene ID: | 10483 |
Gene Name: | Sec23 homolog B, coat complex II component |
Gene Aliases: |
CDA-II, CDAII, CDAN2, CWS7, HEMPAS |
Location: |
Chr.20:18507544-18561415 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC23B | NM_001172745.1 | NP_001166216.1 | ||
| NM_001172746.1 | NP_001166217.1 | |||
| NM_006363.4 | NP_006354.2 | |||
| NM_032985.4 | NP_116780.1 | |||
| NM_032986.3 | NP_116781.1 | |||
| AK296270.1 | ||||
| AK299171.1 | ||||
| AK299513.1 | ||||
| AL540773.3 | ||||
| BC005032.1 | AAH05032.1 | |||
| BC005404.1 | AAH05404.1 | |||
| BC095404.1 | AAH95404.1 | |||
| BM751928.1 | ||||
| BP244573.1 | ||||
| BX398403.2 | ||||
| X97065.1 | CAA65775.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4283n100 | Chr.20:18364565 - 18647377 on Build GRCh38 | Loss |
|
| nsv1062244 | Chr.20:18184227 - 18520274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map