Assay Details
Target Gene Details
Entrez Gene ID: | 2778 |
Gene Name: | GNAS complex locus |
Gene Aliases: |
AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI |
Location: |
Chr.20:58839681-58911196 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAS | NM_001077490.2 | NP_001070958.1 | ||
| NM_001309861.1 | NP_001296790.1 | |||
| NM_001309883.1 | NP_001296812.1 | |||
| NM_016592.3 | NP_057676.1 | |||
| NM_080425.3 | NP_536350.2 | |||
| XM_017027812.1 | XP_016883301.1 | |||
| XM_017027813.1 | XP_016883302.1 | |||
| XM_017027814.1 | XP_016883303.1 | |||
| XM_017027815.1 | XP_016883304.1 | |||
| XM_017027821.1 | XP_016883310.1 | |||
| XM_017027822.1 | XP_016883311.1 | |||
| AF105253.1 | AAF63226.1 | |||
| AJ224867.1 | CAA12164.1 | |||
| AK122771.1 | ||||
| AK315874.1 | ||||
| CN273438.1 | ||||
| CN295544.1 |
Target Gene Details
Entrez Gene ID: | 101927932 |
Gene Name: | uncharacterized LOC101927932 |
Gene Aliases: |
- |
Location: |
Chr.20:58863756-58888810 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927932 | NR_126334.1 | |||
| BI199822.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828763 | Chr.20:58839788 - 58908828 on Build GRCh38 | Loss |
|
| nsv586352 | Chr.20:58846572 - 58904078 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map