Assay Details
Target Gene Details
Entrez Gene ID: | 1591 |
Gene Name: | cytochrome P450 family 24 subfamily A member 1 |
Gene Aliases: |
CP24, CYP24, HCAI, HCINF1, P450-CC24 |
Location: |
Chr.20:54145731-54173985 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP24A1 | NM_000782.4 | 12 | 3005 | NP_000773.2 |
| NM_001128915.1 | 11 | 2807 | NP_001122387.1 | |
| XM_005260304.4 | 12 | 3185 | XP_005260361.1 | |
| XM_017027691.1 | 12 | 3033 | XP_016883180.1 | |
| XM_017027692.1 | XP_016883181.1 | |||
| XM_017027693.1 | 11 | 2987 | XP_016883182.1 | |
| AI400154.1 | 1 | 149 | ||
| AW022349.1 | ||||
| BI491491.1 | 1 | 401 | ||
| L13286.1 | 12 | 2997 | AAA62379.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv586224 | Chr.20:53858311 - 54662951 on Build GRCh38 | Gain |
|
| nsv834010 | Chr.20:54143285 - 54316400 on Build GRCh38 | Loss |
|
| nsv1058882 | Chr.20:54149440 - 54177833 on Build GRCh38 | Loss |
|
| nsv1063981 | Chr.20:54148778 - 54160074 on Build GRCh38 | Loss |
|
| nsv586281 | Chr.20:54148810 - 54154632 on Build GRCh38 | Loss |
|
| esv2762085 | Chr.20:54148790 - 54160086 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map