Assay Details
Target Gene Details
Entrez Gene ID: | 30813 |
Gene Name: | visual system homeobox 1 |
Gene Aliases: |
CAASDS, KTCN, KTCN1, PPCD, PPCD1, PPD, RINX |
Location: |
Chr.20:25069940-25082379 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSX1 | NM_001256271.1 | 4 | 1749 | NP_001243200.1 |
| NM_001256272.1 | 5 | 1930 | NP_001243201.1 | |
| NR_045948.1 | 6 | 2038 | ||
| NR_045951.1 | 5 | 1857 | ||
| XM_017027837.1 | 5 | 2272 | XP_016883326.1 | |
| XM_017027838.1 | 4 | 2091 | XP_016883327.1 | |
| BM680858.1 | 1 | 307 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056345 | Chr.20:24838934 - 25187523 on Build GRCh38 | Gain |
|
| esv3645552 | Chr.20:24494271 - 25093181 on Build GRCh38 | Gain |
|
| nsv522873 | Chr.20:23923336 - 26134874 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map