Assay Details
Target Gene Details
Entrez Gene ID: | 128853 |
Gene Name: | dual specificity phosphatase 15 |
Gene Aliases: |
C20orf57, VHY |
Location: |
Chr.20:31845578-31870744 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUSP15 | NM_001320479.1 | NP_001307408.1 | ||
| XM_017027654.1 | XP_016883143.1 | |||
| XM_017027655.1 | XP_016883144.1 | |||
| XM_017027656.1 | XP_016883145.1 | |||
| XM_017027657.1 | XP_016883146.1 | |||
| XM_017027658.1 | XP_016883147.1 | |||
| XM_017027659.1 | XP_016883148.1 | |||
| XM_017027660.1 | XP_016883149.1 | |||
| XM_017027661.1 | XP_016883150.1 | |||
| XM_017027662.1 | XP_016883151.1 | |||
| AK097430.1 | BAC05048.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833952 | Chr.20:31855501 - 32049500 on Build GRCh38 | Gain |
|
| dgv4300n100 | Chr.20:31247234 - 32033468 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map