Assay Details
Target Gene Details
Entrez Gene ID: | 63939 |
Gene Name: | family with sequence similarity 217 member B |
Gene Aliases: |
C20orf177, dJ551D2.5 |
Location: |
Chr.20:59933764-59948680 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM217B | NM_001190826.1 | NP_001177755.1 | ||
| XM_011528986.2 | XP_011527288.1 | |||
| AK125073.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4336n100 | Chr.20:59857564 - 59943263 on Build GRCh38 | Loss |
|
| nsv1062669 | Chr.20:59878803 - 59940597 on Build GRCh38 | Loss |
|
| nsv1065994 | Chr.20:59845169 - 59951828 on Build GRCh38 | Loss |
|

Genomic Map